Assay Details
Target Gene Details
Entrez Gene ID: | 13846 |
Gene Name: | Eph receptor B4 |
Gene Aliases: |
AI042935, Htk, MDK2, Myk1, Tyro11 |
Location: |
Chr.5:137350109-137374522 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ephb4 | NM_001159571.1 | 16 | 3265 | NP_001153043.1 |
NM_010144.6 | 16 | 3238 | NP_034274.4 | |
AK046884.1 | 16 | 3203 | BAC32908.1 | |
AK049340.1 | 16 | 3239 | BAC33693.1 | |
AK132392.1 | 16 | 3238 | BAE21142.1 | |
BC016505.1 | 16 | 3190 | AAH16505.1 | |
U06834.1 | 17 | 2851 | AAA18591.1 |
More Information
Set Membership: |
Intragenic Exonic |