Assay Details
Target Gene Details
Entrez Gene ID: | 13846 |
Gene Name: | Eph receptor B4 |
Gene Aliases: |
AI042935, Htk, MDK2, Myk1, Tyro11 |
Location: |
Chr.5:137350109-137374522 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ephb4 | NM_001159571.1 | NP_001153043.1 | ||
NM_010144.6 | NP_034274.4 | |||
AK046884.1 | BAC32908.1 | |||
AK049340.1 | BAC33693.1 | |||
AK132392.1 | BAE21142.1 | |||
AK199938.1 | ||||
BC016505.1 | AAH16505.1 | |||
U06834.1 | AAA18591.1 |
More Information
Set Membership: |
Intragenic Non-exonic |