Assay Details
Target Gene Details
Entrez Gene ID: | 67848 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 55 |
Gene Aliases: |
2810021H22Rik, mKIAA1595 |
Location: |
Chr.5:124552544-124569660 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx55 | NM_001190795.1 | NP_001177724.1 | ||
NM_026409.4 | NP_080685.2 | |||
AK012782.1 | BAB28466.1 | |||
AK017554.1 | BAB30802.1 | |||
AK033741.1 | BAC28459.1 | |||
AK077525.1 | BAC36845.1 | |||
AK129402.1 | BAC98212.1 | |||
AK154422.1 | BAE32574.1 | |||
AK167149.1 | 3 | 268 | ||
BC043052.1 | AAH43052.1 | |||
BC117786.1 | ||||
BC117787.1 | ||||
BY202366.1 |