Assay Details
Target Gene Details
Entrez Gene ID: | 140904 |
Gene Name: | calneuron 1 |
Gene Aliases: |
9630012C17Rik, Cabp8, MNCb-0849 |
Location: |
Chr.5:130369356-130846325 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Caln1 | NM_021371.2 | NP_067346.1 | ||
NM_181045.1 | NP_851388.1 | |||
AB041539.1 | BAA95024.1 | |||
AF282251.1 | AAK15156.1 | |||
AK035865.1 | BAC29219.1 | |||
AK081011.1 | BAC38115.1 | |||
AK158641.1 | BAE34592.1 | |||
AK158892.1 | BAE34716.1 | |||
AK163648.1 | BAE37437.1 | |||
BC057129.1 | AAH57129.1 | |||
BC058963.1 | AAH58963.1 |