Assay Details
Target Gene Details
Entrez Gene ID: | 208890 |
Gene Name: | solute carrier family 26, member 7 |
Gene Aliases: |
- |
Location: |
Chr.4:14502430-14621971 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc26a7 | NM_145947.2 | NP_666059.2 | ||
AF345194.1 | AAO49172.1 | |||
AK054327.1 | ||||
BC026928.1 | AAH26928.1 |
More Information
Set Membership: |
Intragenic Non-exonic |