Assay Details
Target Gene Details
Entrez Gene ID: | 69934 |
Gene Name: | tRNA methyltransferase 10B |
Gene Aliases: |
2610042J10Rik, Rg9mtd3, Rnmtd3 |
Location: |
Chr.4:45297071-45316131 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trmt10b | NM_027266.4 | 2 | 94 | NP_081542.2 |
AK011746.1 | 2 | 107 | BAB27815.1 | |
AK030465.1 | 2 | 95 | BAC26974.1 | |
BC019428.1 | 1 | 71 | AAH19428.1 |
More Information
Set Membership: |
Intragenic Exonic |