Assay Details
Target Gene Details
Entrez Gene ID: | 230233 |
Gene Name: | inhibitor of kappa light polypeptide enhancer in B cells, kinase complex-associated protein |
Gene Aliases: |
3110040G09Rik, 6030413P05, C78473, Elp1, IKAP |
Location: |
Chr.4:56749680-56802331 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 25 - Exon 26 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ikbkap | NM_026079.3 | NP_080355.2 | ||
AF140786.1 | AAO15309.1 | |||
AF367244.1 | AAL36025.1 | |||
AF387811.1 | AAL40925.1 | |||
AK031367.1 | ||||
AK035944.1 | BAC29252.1 | |||
AK147153.1 | BAE27720.1 | |||
AK165168.1 | ||||
BC052387.1 | AAH52387.2 |
More Information
Set Membership: |
Intragenic Non-exonic |