Assay Details
Target Gene Details
Entrez Gene ID: | 14915 |
Gene Name: | guanylate cyclase activator 2a (guanylin) |
Gene Aliases: |
Guca2 |
Location: |
Chr.4:119637732-119639465 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Guca2a | NM_008190.1 | 2 | 156 | NP_032216.1 |
AK075814.1 | 2 | 175 | BAC35981.1 | |
BC012640.1 | 2 | 167 | AAH12640.1 | |
L05516.1 | 2 | 150 | AAA37715.1 | |
M95175.1 | 2 | 156 | AAA37758.1 |
More Information
Set Membership: |
Intragenic Exonic |