Assay Details
Target Gene Details
Entrez Gene ID: | 76238 |
Gene Name: | glyoxylate reductase/hydroxypyruvate reductase |
Gene Aliases: |
1110059D05Rik, 6430629L09Rik |
Location: |
Chr.4:44981387-44992085 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Grhpr | NM_080289.1 | NP_525028.1 | ||
AK004307.1 | ||||
AK018293.1 | ||||
AK168351.1 | BAE40288.1 | |||
AK172201.1 | BAE42878.1 | |||
AY113690.1 | AAM52985.1 | |||
BC010194.1 | AAH10194.1 |
More Information
Set Membership: |
Intragenic Non-exonic |