Assay Details
Target Gene Details
Entrez Gene ID: | 72033 |
Gene Name: | TSC22 domain family, member 2 |
Gene Aliases: |
1810043J12Rik, 5530402M19Rik |
Location: |
Chr.3:58415531-58466787 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tsc22d2 | NM_001081229.1 | NP_001074698.1 | ||
AK007760.1 | BAB25238.2 | |||
AK156074.1 | BAE33573.1 | |||
BC058221.1 | AAH58221.1 | |||
BC117998.1 | ||||
EU004151.1 |