Assay Details
Target Gene Details
Entrez Gene ID: | 26570 |
Gene Name: | solute carrier family 7 (cationic amino acid transporter, y+ system), member 11 |
Gene Aliases: |
9930009M05Rik, AI451155, sut, xCT |
Location: |
Chr.3:50364936-50499087 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc7a11 | NM_011990.2 | 11 | 1663 | NP_036120.1 |
AB022345.1 | 11 | 1630 | BAA77220.1 | |
AK076322.1 | 11 | 1638 | ||
AK089202.1 | 11 | 1636 | BAC40792.1 | |
AK162233.1 | 11 | 1438 | ||
AK163168.1 | 11 | 1637 | BAE37218.1 | |
AK165599.1 | 11 | 1639 | BAE38284.1 | |
AK171192.1 | 11 | 1661 | BAE42302.1 | |
AK171252.1 | 11 | 1637 | BAE42344.1 | |
AY766236.1 | 11 | 1630 | AAX10866.1 | |
BC141402.1 | 11 | 1560 | ||
BC141408.1 | 11 | 1560 |
More Information
Set Membership: |
Intragenic Exonic |