Assay Details
Target Gene Details
Entrez Gene ID: | 209232 |
Gene Name: | WAP four-disulfide core domain 5 |
Gene Aliases: |
BC019734, Prg5, Wap1 |
Location: |
Chr.2:164176325-164182742 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wfdc5 | NM_145369.3 | NP_663344.2 | ||
AK075896.1 | ||||
AK131905.1 | BAE20863.1 | |||
AK131906.1 | BAE20864.1 | |||
AY669505.1 | AAU89495.1 | |||
BC019734.1 | AAH19734.1 |
More Information
Set Membership: |
Intragenic Non-exonic |