Assay Details
Target Gene Details
Entrez Gene ID: | 320106 |
Gene Name: | solute carrier family 38, member 11 |
Gene Aliases: |
9330158F14Rik |
Location: |
Chr.2:65315799-65364394 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc38a11 | NM_177074.2 | 4 | 421 | NP_796048.2 |
AK034126.1 | 4 | 430 | BAC28596.1 | |
AK041015.1 | 4 | 319 | BAC30781.1 | |
AK043610.1 | 4 | 430 | ||
AK139966.1 | 4 | 371 | BAE24200.1 | |
CJ055756.1 |
More Information
Set Membership: |
Intragenic Exonic |