Assay Details
Target Gene Details
Entrez Gene ID: | 320558 |
Gene Name: | synaptonemal complex protein 2 |
Gene Aliases: |
3830402K23Rik, 4930518F03Rik |
Location: |
Chr.2:178345295-178407658 on Build GRCm38 |
Assay Gene Location: | Within Exon 34 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sycp2 | NM_177191.3 | 34 | 3502 | NP_796165.2 |
AK029735.1 | 7 | 813 | BAC26589.1 | |
AK036624.1 | 12 | 1350 | ||
DQ103262.1 | 34 | 3502 |
More Information
Set Membership: |
Intragenic Exonic |