Assay Details
Target Gene Details
Entrez Gene ID: | 20779 |
Gene Name: | Rous sarcoma oncogene |
Gene Aliases: |
AW259666, pp60c-src |
Location: |
Chr.2:157423901-157471838 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Src | NM_001025395.2 | 5 | 684 | NP_001020566.1 |
NM_009271.3 | 5 | 684 | NP_033297.2 | |
BC039953.1 | 5 | 694 | AAH39953.1 | |
CX218263.1 | 2 | 183 | ||
M17031.1 | 2 | 300 | AAA40135.1 |