Assay Details
Target Gene Details
Entrez Gene ID: | 629499 |
Gene Name: | maestro heat-like repeat family member 8 |
Gene Aliases: |
4922505G16Rik |
Location: |
Chr.2:157208548-157279549 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mroh8 | NM_001039557.4 | NP_001034646.2 | ||
AK029507.1 | ||||
AK029675.1 | 1 | 335 | BAC26558.1 | |
AK089157.1 | BAC40769.1 | |||
AK132763.1 | BAE21344.1 | |||
BC150937.1 |
Target Gene Details
Entrez Gene ID: | 20014 |
Gene Name: | ribophorin II |
Gene Aliases: |
1300012C06Rik, AV261018, Rpn-2 |
Location: |
Chr.2:157279011-157326318 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rpn2 | NM_019642.4 | 1 | 119 | NP_062616.2 |
AK004968.1 | 1 | 120 | BAB23707.1 | |
AK018761.2 | 1 | 158 | ||
BC046806.1 | 1 | 147 | AAH46806.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |