Assay Details
Target Gene Details
Entrez Gene ID: | 75526 |
Gene Name: | epididymal peptidase inhibitor |
Gene Aliases: |
1700024E17Rik, Spinlw1 |
Location: |
Chr.2:164588343-164593571 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eppin | NM_029325.2 | 1 | 112 | NP_083601.1 |
AF346413.2 | 1 | 135 | AAK31335.1 | |
AK006296.1 | 1 | 114 | BAB24514.1 | |
BC048637.1 | 1 | 135 | AAH48637.1 |