Assay Details
Target Gene Details
Entrez Gene ID: | 57329 |
Gene Name: | otoraplin |
Gene Aliases: |
CDRAP, Fdp, MIA, MIAL |
Location: |
Chr.2:143078492-143081700 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Otor | NM_020595.2 | NP_065620.1 | ||
AF243504.1 | AAG42355.1 | |||
AJ243939.1 | CAC27444.1 | |||
AK047965.1 | 1 | 569 | ||
BC139152.1 | ||||
BC139153.1 |