Assay Details
Target Gene Details
Entrez Gene ID: | 17932 |
Gene Name: | myelin transcription factor 1 |
Gene Aliases: |
Nzf2, Nzf2a, Nzf2b, Nztf2 |
Location: |
Chr.2:181763332-181827781 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Myt1 | NM_001171615.1 | 11 | 1968 | NP_001165086.1 |
NM_001171616.1 | 10 | 2078 | NP_001165087.1 | |
NM_001171680.1 | 5 | 2158 | NP_001165151.1 | |
NM_008665.4 | 11 | 2192 | NP_032691.2 | |
AB082378.1 | 11 | 2179 | BAC16512.1 | |
AB093267.2 | 10 | 1785 | BAC41451.2 | |
AF004294.1 | 5 | 2159 | AAC53456.1 | |
AK133358.1 | 5 | 889 | ||
AK135131.1 | 3 | 354 | BAE22433.1 | |
BC063252.1 | 11 | 1968 | AAH63252.1 |
More Information
Set Membership: |
Intragenic Exonic |