Assay Details
Target Gene Details
Entrez Gene ID: | 76467 |
Gene Name: | methionine sulfoxide reductase B2 |
Gene Aliases: |
2310050L06Rik, Mrsb, Msrb, Pilb |
Location: |
Chr.2:19371636-19394976 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Msrb2 | NM_029619.2 | 2 | 190 | NP_083895.1 |
AK009912.1 | 2 | 181 | ||
BC021619.1 | 2 | 190 | AAH21619.1 |
More Information
Set Membership: |
Intragenic Exonic |