Assay Details
Target Gene Details
Entrez Gene ID: | 66634 |
Gene Name: | minichromosome maintenance deficient 8 (S. cerevisiae) |
Gene Aliases: |
5730432L01Rik |
Location: |
Chr.2:132816141-132844197 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mcm8 | NM_001291054.1 | 5 | 776 | NP_001277983.1 |
NM_025676.4 | 4 | 692 | NP_079952.2 | |
AK017607.1 | 5 | 674 | ||
AK133858.1 | 5 | 733 | BAE21892.1 | |
BC046780.1 | 5 | 723 | AAH46780.1 | |
BC052070.1 | 4 | 498 | AAH52070.1 |
More Information
Set Membership: |
Intragenic Exonic |