Assay Details
Target Gene Details
Entrez Gene ID: | 16558 |
Gene Name: | kinesin family member 16B |
Gene Aliases: |
8430434E15Rik, AA623607, AI789011, C80253, C80902, mKIAA1590 |
Location: |
Chr.2:142618334-142901595 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kif16b | NM_001081133.2 | 20 | 3354 | NP_001074602.1 |
AB610968.1 | 21 | 3442 | ||
AK129401.1 | 8 | 1558 | BAC98211.1 | |
AK142909.1 | 2 | 654 | ||
AK160835.1 | 4 | 1574 | BAE36039.1 |
More Information
Set Membership: |
Intragenic Exonic |