Assay Details
Target Gene Details
Entrez Gene ID: | 75608 |
Gene Name: | charged multivesicular body protein 4B |
Gene Aliases: |
2010012F05Rik, C76846, Snf7-2 |
Location: |
Chr.2:154657026-154694783 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Chmp4b | NM_029362.3 | NP_083638.1 | ||
AK008205.1 | ||||
AK156473.1 | BAE33724.1 | |||
AK159193.1 | BAE34888.1 | |||
AK196942.1 | ||||
BC006905.1 | AAH06905.1 | |||
BC011429.1 | AAH11429.1 | |||
BC059279.1 | AAH59279.1 |
More Information
Set Membership: |
Intragenic Non-exonic |