Assay Details
Target Gene Details
Entrez Gene ID: | 12613 |
Gene Name: | carboxyl ester lipase |
Gene Aliases: |
1810036E18Rik, BAL |
Location: |
Chr.2:28555819-28563362 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cel | NM_009885.1 | 8 | 1033 | NP_034015.1 |
AK131818.1 | 8 | 1051 | BAE20818.1 | |
AK131824.1 | 8 | 1051 | BAE20820.1 | |
BC006872.1 | 8 | 1045 | AAH06872.1 | |
U33169.1 | 8 | 1033 | AAA92088.1 | |
U37386.1 | 8 | 1037 | AAC52279.1 |
More Information
Set Membership: |
Intragenic Exonic |