Assay Details
Target Gene Details
Entrez Gene ID: | 227526 |
Gene Name: | cerebral dopamine neurotrophic factor |
Gene Aliases: |
9330140G23, Armetl1 |
Location: |
Chr.2:3512966-3526376 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdnf | NM_177647.4 | 2 | 243 | NP_808315.1 |
AK034009.1 | 2 | 245 | BAC28545.1 | |
BC110560.2 | 2 | 231 | AAI10561.1 |