Assay Details
Target Gene Details
Entrez Gene ID: | 19275 |
Gene Name: | protein tyrosine phosphatase, receptor type, N |
Gene Aliases: |
IA-2, mIA-A, mKIAA4064 |
Location: |
Chr.1:75247035-75264497 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptprn | NM_008985.2 | 13 | 1781 | NP_033011.2 |
AF332083.1 | 10 | 1385 | AAK56111.1 | |
AF332084.1 | 10 | 1376 | AAK56112.1 | |
AK041296.1 | 13 | 1787 | ||
AK076223.1 | 13 | 1762 | ||
AK139398.1 | 12 | 2301 | BAE23994.1 | |
AK220467.1 | 13 | 1739 | BAD90494.1 | |
BC068165.1 | 13 | 1713 | AAH68165.1 | |
DQ832283.1 | 13 | 1758 | ||
U11812.1 | 13 | 1699 | AAA52102.1 | |
X74438.1 | 13 | 1798 | CAA52453.1 |
More Information
Set Membership: |
Intragenic Exonic |