Assay Details
Target Gene Details
Entrez Gene ID: | 215378 |
Gene Name: | bone morphogenetic protein/retinoic acid inducible neural specific 3 |
Gene Aliases: |
B830045N13Rik, Fam5c |
Location: |
Chr.1:146495666-146902474 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Brinp3 | NM_001145807.1 | 7 | 1213 | NP_001139279.1 |
NM_153539.3 | 7 | 1571 | NP_705767.3 | |
AK046878.1 | 7 | 1572 | ||
AK083233.1 | 8 | 1262 | ||
BC030498.1 | 6 | 1189 | AAH30498.1 | |
BC099956.1 | 7 | 1548 | AAH99956.1 |