Assay Details
Target Gene Details
Entrez Gene ID: | 22596 |
Gene Name: | X-ray repair complementing defective repair in Chinese hamster cells 5 |
Gene Aliases: |
AI314015, Ku80, Ku86 |
Location: |
Chr.1:72307417-72394953 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Xrcc5 | NM_009533.2 | NP_033559.2 | ||
AF166486.1 | AAD49720.1 | |||
AK010416.1 | BAB26920.3 | |||
AK081633.1 | BAC38276.1 | |||
AK089944.1 | ||||
AK165470.1 | BAE38207.1 | |||
AK167312.1 | BAE39415.1 | |||
AK168913.1 | BAE40726.1 | |||
AK169838.1 | BAE41402.1 | |||
BC029218.1 | AAH29218.1 | |||
BC051660.1 | AAH51660.1 | |||
X66323.1 | CAA46999.1 |
More Information
Set Membership: |
Intragenic Non-exonic |