Assay Details
Target Gene Details
Entrez Gene ID: | 227059 |
Gene Name: | solute carrier family 39 (zinc transporter), member 10 |
Gene Aliases: |
2900042E17Rik, 5430433I10, mKIAA1265 |
Location: |
Chr.1:46807544-46925772 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc39a10 | NM_172653.2 | 4 | 1319 | NP_766241.2 |
AK049099.1 | 2 | 65 | BAC33542.1 | |
AK122483.1 | 3 | 753 | BAC65765.1 | |
AK136117.1 | 3 | 1031 | ||
BC052880.1 | 4 | 1433 | AAH52880.1 | |
BC059214.1 | 2 | 203 | AAH59214.1 | |
BC062918.1 | 4 | 1319 | AAH62918.1 |
More Information
Set Membership: |
Intragenic Exonic |