Assay Details
Target Gene Details
Entrez Gene ID: | 320718 |
Gene Name: | solute carrier family 26, member 9 |
Gene Aliases: |
E030002L01Rik |
Location: |
Chr.1:131744022-131771504 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc26a9 | NM_177243.4 | 3 | 302 | NP_796217.2 |
AK078966.1 | 3 | 303 | BAC37482.1 | |
AK086815.1 | 3 | 303 | BAC39749.1 | |
AY034145.1 | 2 | 215 | AAK54448.1 |
More Information
Set Membership: |
Intragenic Exonic |