Assay Details
Target Gene Details
Entrez Gene ID: | 24012 |
Gene Name: | regulator of G protein signaling 7 |
Gene Aliases: |
- |
Location: |
Chr.1:175059076-175493238 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rgs7 | NM_001199003.1 | NP_001185932.1 | ||
NM_011880.3 | NP_036010.2 | |||
AF011360.1 | AAC99483.1 | |||
BC051133.1 | AAH51133.1 | |||
CJ060456.1 | ||||
CJ060511.1 |