Assay Details
Target Gene Details
Entrez Gene ID: | 56210 |
Gene Name: | REV1 homolog (S. cerevisiae) |
Gene Aliases: |
1110027I23Rik, AU022044, Rev1l |
Location: |
Chr.1:38052786-38129919 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rev1 | NM_019570.3 | 11 | 1955 | NP_062516.2 |
AB057418.1 | 10 | 1744 | BAB64933.1 | |
AF179302.1 | 10 | 1744 | AAF23323.1 | |
AK048838.1 | 11 | 1960 | ||
BC058093.1 | 11 | 1955 | AAH58093.1 |
More Information
Set Membership: |
Intragenic Exonic |