Assay Details
Target Gene Details
Entrez Gene ID: | 70247 |
Gene Name: | proteasome (prosome, macropain) 26S subunit, non-ATPase, 1 |
Gene Aliases: |
2410026J11Rik, P112, S1 |
Location: |
Chr.1:86064619-86139295 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 19 - Intron 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Psmd1 | NM_027357.2 | NP_081633.1 | ||
AK010596.1 | ||||
AK087947.1 | BAC40055.2 | |||
AK153386.1 | BAE31950.1 | |||
AK159123.1 | BAE34838.1 | |||
AK159750.1 | ||||
AK180030.1 | ||||
AK202172.1 | ||||
AK219841.1 | ||||
BC018172.1 | ||||
BC022955.1 | ||||
BC025802.1 | ||||
BC031587.1 | AAH31587.1 | |||
BC039937.1 | AAH39937.1 | |||
BC089028.1 | AAH89028.1 | |||
BC138526.1 | ||||
BC138527.1 |
More Information
Set Membership: |
Intragenic Non-exonic |