Assay Details
Target Gene Details
Entrez Gene ID: | 70247 |
Gene Name: | proteasome (prosome, macropain) 26S subunit, non-ATPase, 1 |
Gene Aliases: |
2410026J11Rik, P112, S1 |
Location: |
Chr.1:86064619-86139295 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Psmd1 | NM_027357.2 | 14 | 1835 | NP_081633.1 |
AK087947.1 | 14 | 1786 | BAC40055.2 | |
AK153386.1 | 14 | 1836 | BAE31950.1 | |
AK159123.1 | 14 | 1835 | BAE34838.1 | |
AK159750.1 | 14 | 1814 | ||
BC022955.1 | 14 | 1725 | ||
BC025802.1 | 14 | 1739 | ||
BC031587.1 | 14 | 1766 | AAH31587.1 | |
BC039937.1 | 14 | 1812 | AAH39937.1 | |
BC089028.1 | 14 | 1782 | AAH89028.1 | |
BC138526.1 | 14 | 1798 | ||
BC138527.1 | 14 | 1798 |
More Information
Set Membership: |
Intragenic Exonic |