Assay Details
Target Gene Details
Entrez Gene ID: | 15559 |
Gene Name: | 5-hydroxytryptamine (serotonin) receptor 2B |
Gene Aliases: |
5-HT2B, AJ012488, AV377389 |
Location: |
Chr.1:86097546-86113662 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Htr2b | NM_008311.2 | 3 | 818 | NP_032337.2 |
AF498254.1 | 2 | 491 | AAM22971.1 | |
AF498255.1 | 2 | 491 | AAM22972.1 | |
AK033713.1 | 3 | 818 | BAC28441.1 | |
AY131263.1 | ||||
AY131264.1 | 2 | 281 | AAN37496.1 | |
BC023690.1 | 3 | 767 | AAH23690.1 | |
Z15119.1 | 2 | 526 | CAA78824.1 |
Target Gene Details
Entrez Gene ID: | 70247 |
Gene Name: | proteasome (prosome, macropain) 26S subunit, non-ATPase, 1 |
Gene Aliases: |
2410026J11Rik, P112, S1 |
Location: |
Chr.1:86064619-86139295 on Build GRCm38 |
Assay Gene Location: | Within Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Psmd1 | NM_027357.2 | NP_081633.1 | ||
AK087947.1 | BAC40055.2 | |||
AK153386.1 | BAE31950.1 | |||
AK159123.1 | BAE34838.1 | |||
AK159750.1 | ||||
BC018172.1 | ||||
BC022955.1 | ||||
BC025802.1 | ||||
BC031587.1 | AAH31587.1 | |||
BC039937.1 | AAH39937.1 | |||
BC089028.1 | AAH89028.1 | |||
BC138526.1 | ||||
BC138527.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |