Assay Details
Target Gene Details
Entrez Gene ID: | 226982 |
Gene Name: | eukaryotic translation initiation factor 5B |
Gene Aliases: |
A030003E17Rik, BC018347, IF2 |
Location: |
Chr.1:37998000-38055579 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif5b | NM_198303.2 | NP_938045.2 | ||
AK145146.1 | BAE26259.1 | |||
AK162176.1 | ||||
CN724918.1 |
More Information
Set Membership: |
Intragenic Non-exonic |