Assay Details
Target Gene Details
Entrez Gene ID: | 72949 |
Gene Name: | cyclin T2 |
Gene Aliases: |
2900041I18Rik, C81304, CycT2, CycT2a, CycT2b |
Location: |
Chr.1:127774164-127804837 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccnt2 | NM_028399.1 | NP_082675.1 | ||
AK013634.1 | BAB28933.1 | |||
AK049863.1 | ||||
AK152399.1 | BAE31187.1 | |||
AK154439.1 | BAE32588.1 | |||
AK156621.1 | ||||
BC054122.1 | AAH54122.1 | |||
JX965957.1 | ||||
JX965958.1 |
More Information
Set Membership: |
Intragenic Non-exonic |