Assay Details
Target Gene Details
Entrez Gene ID: | 74104 |
Gene Name: | ATP-binding cassette, sub-family B (MDR/TAP), member 6 |
Gene Aliases: |
1200005B17Rik |
Location: |
Chr.1:75171640-75180283 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abcb6 | NM_023732.2 | 2 | 786 | NP_076221.1 |
AF045562.1 | 2 | 647 | AAC05261.1 | |
AK004605.1 | 2 | 866 | BAB23404.1 | |
AK040323.1 | 2 | 790 | ||
AK150853.1 | 2 | 874 | BAE29909.1 | |
AK151165.1 | 2 | 895 | BAE30168.1 | |
AK152584.1 | 2 | 874 | BAE31334.1 | |
AK164003.1 | 2 | 790 | ||
AK168642.1 | 2 | 887 | BAE40501.1 | |
BC006634.1 | 2 | 786 | AAH06634.1 |
More Information
Set Membership: |
Intragenic Exonic |