Assay Details
Target Gene Details
Entrez Gene ID: | 19275 |
Gene Name: | protein tyrosine phosphatase, receptor type, N |
Gene Aliases: |
IA-2, mIA-A, mKIAA4064 |
Location: |
Chr.1:75247035-75264497 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptprn | NM_008985.2 | NP_033011.2 | ||
AF332083.1 | AAK56111.1 | |||
AF332084.1 | AAK56112.1 | |||
AK041296.1 | ||||
AK076223.1 | ||||
AK139398.1 | BAE23994.1 | |||
AK220467.1 | BAD90494.1 | |||
BC068165.1 | AAH68165.1 | |||
DQ832283.1 | ||||
U11812.1 | AAA52102.1 | |||
X74438.1 | CAA52453.1 |
More Information
Set Membership: |
Intragenic Non-exonic |