Assay Details
Target Gene Details
Entrez Gene ID: | 76709 |
Gene Name: | actin related protein 2/3 complex, subunit 2 |
Gene Aliases: |
2210023N03Rik, 34kDa, p34-Arc |
Location: |
Chr.1:74236504-74268213 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arpc2 | NM_029711.1 | NP_083987.1 | ||
AK008777.1 | BAB25889.2 | |||
AK083263.1 | ||||
AK151513.1 | BAE30463.1 | |||
AK152356.1 | BAE31148.1 | |||
AK170061.1 | ||||
AK211266.1 | ||||
BC099504.1 | AAH99504.1 | |||
BC115750.1 | AAI15751.1 |