Assay Details
Target Gene Details
Entrez Gene ID: | 23560 |
Gene Name: | GTP binding protein 4 |
Gene Aliases: |
CRFG, NGB, NOG1 |
Location: |
Chr.10:988409-1017768 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GTPBP4 | NM_012341.2 | NP_036473.2 | ||
AF120334.1 | AAD09830.1 | |||
AF325353.1 | AAK13444.1 | |||
AK001548.1 | ||||
AK001552.1 | BAA91752.1 | |||
AK097093.1 | ||||
AK222861.1 | BAD96581.1 | |||
AK295231.1 | ||||
AK301721.1 | ||||
AK302219.1 | ||||
BC033784.1 | AAH33784.1 | |||
BC038975.2 | AAH38975.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2729239 | Chr.10:1009843 - 1010905 on Build GRCh38 | Deletion | GTPBP4 |
esv2660097 | Chr.10:526001 - 1246069 on Build GRCh38 | Deletion | ADARB2 LARP4B LOC105376341 LOC101927762 GTPBP4 LOC105376339 IDI2 LOC101930421 LINC00200 PRR26 IDI1 IDI2-AS1 DIP2C WDR37 MIR5699 |
esv2551854 | Chr.10:1009527 - 1010525 on Build GRCh38 | Insertion | GTPBP4 |
esv24871 | Chr.10:1010011 - 1011435 on Build GRCh38 | Gain | GTPBP4 |
esv3891735 | Chr.10:978274 - 1064524 on Build GRCh38 | Gain | IDI2 GTPBP4 IDI1 IDI2-AS1 WDR37 |
nsv549765 | Chr.10:688289 - 1083587 on Build GRCh38 | Gain | IDI2 LARP4B LOC105376341 LOC101927762 GTPBP4 IDI1 IDI2-AS1 DIP2C WDR37 |
esv2729251 | Chr.10:1009847 - 1011600 on Build GRCh38 | Deletion | GTPBP4 |
nsv1161921 | Chr.10:823011 - 1033401 on Build GRCh38 | Duplication | IDI2 LARP4B LOC105376341 LOC101927762 GTPBP4 IDI2-AS1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |