Assay Details
Target Gene Details
Entrez Gene ID: | 55816 |
Gene Name: | docking protein 5 |
Gene Aliases: |
C20orf180, IRS-6, IRS6 |
Location: |
Chr.20:54475472-54651171 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DOK5 | NM_018431.4 | NP_060901.2 | ||
NM_177959.2 | NP_808874.1 | |||
XM_011528904.1 | XP_011527206.1 | |||
AF132732.1 | AAF66443.1 | |||
AF466368.1 | AAL74194.1 | |||
AK313222.1 | ||||
BC008992.1 | AAH08992.1 | |||
BI758267.1 | ||||
DA161365.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv586224 | Chr.20:53858311 - 54662951 on Build GRCh38 | Gain | SUMO1P1 CYP24A1 BCAS1 PFDN4 MIR4756 DOK5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |