Assay Details
Target Gene Details
Entrez Gene ID: | 259240 |
Gene Name: | WAP four-disulfide core domain 9 |
Gene Aliases: |
WAP9, dJ688G8.2 |
Location: |
Chr.20:45607939-45631268 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WFDC9 | NM_147198.3 | NP_671731.1 | ||
AY047610.1 | AAK97772.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv586081 | Chr.20:45573240 - 45747557 on Build GRCh38 | Loss | WFDC10A WFDC11 WFDC9 WFDC13 WFDC8 MIR3617 WFDC3 SPINT4 WFDC10B |
nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss | MATN4 SLPI TP53TG5 WISP2 WFDC9 PABPC1L WFDC8 STK4-AS1 PIGT WFDC12 RBPJL SDC4 SYS1 SPINT3 RIMS4 PI3 MIR6812 YWHAB WFDC5 DBNDD2 STK4 WFDC6 KCNK15 SEMG2 EPPIN-WFDC6 WFDC2 SYS1-DBNDD2 TOMM34 WFDC10A EPPIN KCNS1 SEMG1 KCNK15-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |