Assay Details
Target Gene Details
Entrez Gene ID: | 8725 |
Gene Name: | URI1, prefoldin like chaperone |
Gene Aliases: |
C19orf2, NNX3, PPP1R19, RMP, URI |
Location: |
Chr.19:29923644-30016612 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
URI1 | NM_001252641.1 | NP_001239570.1 | ||
NM_003796.3 | NP_003787.2 | |||
NR_045557.1 | ||||
XM_005259362.2 | XP_005259419.1 | |||
XM_005259363.4 | XP_005259420.1 | |||
XM_011527435.1 | XP_011525737.1 | |||
AF091095.1 | AAD08679.1 | |||
AK001437.1 | ||||
AK226173.1 | ||||
AK292170.1 | ||||
AK314313.1 | ||||
BC026184.2 | AAH26184.2 | |||
BC067259.1 | AAH67259.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1063678 | Chr.19:29937920 - 29987524 on Build GRCh38 | Loss | URI1 |
dgv3510n100 | Chr.19:29961142 - 30020645 on Build GRCh38 | Loss | URI1 |
nsv579282 | Chr.19:29960594 - 30000137 on Build GRCh38 | Loss | URI1 |
dgv3511n100 | Chr.19:29964373 - 29988391 on Build GRCh38 | Loss | URI1 |
esv3644130 | Chr.19:29966199 - 29973554 on Build GRCh38 | Loss | URI1 |
dgv3509n100 | Chr.19:29954152 - 30028098 on Build GRCh38 | Loss | URI1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |