Assay Details
Target Gene Details
Entrez Gene ID: | 11140 |
Gene Name: | cell division cycle 37 |
Gene Aliases: |
P50CDC37 |
Location: |
Chr.19:10391128-10403595 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDC37 | NM_007065.3 | NP_008996.1 | ||
XM_011527652.2 | XP_011525954.1 | |||
BC000083.2 | AAH00083.1 | |||
BC008793.2 | AAH08793.1 | |||
BE796008.1 | ||||
BT006796.1 | AAP35442.1 | |||
CR542249.1 | CAG47045.1 | |||
U43077.1 | AAB63979.1 | |||
U63131.1 | AAB04798.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv518008 | Chr.19:10338682 - 10426592 on Build GRCh38 | Gain | PDE4A MIR1181 TYK2 ICAM3 CDC37 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |