Assay Details
Target Gene Details
Entrez Gene ID: | 58510 |
Gene Name: | proline dehydrogenase 2 |
Gene Aliases: |
HSPOX1, HYPDH |
Location: |
Chr.19:35799987-35813299 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRODH2 | NM_021232.1 | NP_067055.1 | ||
XM_017027084.1 | XP_016882573.1 | |||
XM_017027085.1 | XP_016882574.1 | |||
U80018.1 | AAF21465.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953286 | Chr.19:35747000 - 35868398 on Build GRCh38 | Deletion | KIRREL2 NPHS1 PRODH2 LINC01529 ARHGAP33 PROSER3 LIN37 PSENEN HSPB6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |