Assay Details
Target Gene Details
Entrez Gene ID: | 11338 |
Gene Name: | U2 small nuclear RNA auxiliary factor 2 |
Gene Aliases: |
U2AF65 |
Location: |
Chr.19:55654050-55674716 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
U2AF2 | NM_001012478.1 | NP_001012496.1 | ||
NM_007279.2 | NP_009210.1 | |||
XM_006722994.2 | XP_006723057.1 | |||
XM_011526410.1 | XP_011524712.1 | |||
AB451261.1 | ||||
AB451387.1 | ||||
BC008740.2 | AAH08740.1 | |||
BC030574.2 | AAH30574.1 | |||
X64044.1 | CAA45409.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1061024 | Chr.19:55655735 - 55705993 on Build GRCh38 | Gain | U2AF2 EPN1 |
nsv953625 | Chr.19:55637035 - 55699834 on Build GRCh38 | Deletion | CCDC106 ZNF581 U2AF2 ZNF580 LOC107983998 EPN1 |
nsv833882 | Chr.19:55647267 - 55815198 on Build GRCh38 | Loss | CCDC106 U2AF2 NLRP11 NLRP9 RFPL4AL1 RFPL4A EPN1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |