Assay Details
Target Gene Details
Entrez Gene ID: | 55015 |
Gene Name: | pre-mRNA processing factor 39 |
Gene Aliases: |
- |
Location: |
Chr.14:45084099-45115601 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRPF39 | NM_017922.3 | NP_060392.3 | ||
AK000673.1 | BAA91318.1 | |||
AK307530.1 | ||||
AK308415.1 | ||||
AL599626.1 | ||||
BC125126.1 | ||||
BC125127.1 | ||||
DC306976.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832785 | Chr.14:45051467 - 45241538 on Build GRCh38 | Loss | LOC105370474 FAM179B FANCM PRPF39 FKBP3 MIS18BP1 SNORD127 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |