Assay Details
Target Gene Details
Entrez Gene ID: | 23428 |
Gene Name: | solute carrier family 7 member 8 |
Gene Aliases: |
LAT2, LPI-PC1 |
Location: |
Chr.14:23125295-23183660 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC7A8 | NM_001267036.1 | NP_001253965.1 | ||
NM_001267037.1 | NP_001253966.1 | |||
NM_012244.3 | NP_036376.2 | |||
NM_182728.2 | NP_877392.1 | |||
NR_049767.1 | ||||
AB037669.1 | BAB21519.1 | |||
AF087908.1 | AAP97206.1 | |||
AF087911.1 | AAP97208.1 | |||
AF135828.1 | AAF05695.1 | |||
AF171669.1 | AAF20381.1 | |||
AK223610.1 | BAD97330.1 | |||
AK296702.1 | ||||
AK300384.1 | ||||
AK307609.1 | ||||
AK313465.1 | ||||
AL365342.1 | ||||
AL365343.2 | ||||
AL365346.1 | ||||
AL365414.1 | ||||
BC036825.1 | AAH36825.1 | |||
BC052250.1 | AAH52250.1 | |||
BX248288.1 | CAD62616.1 | |||
DC348596.1 | ||||
Y18483.1 | CAB40137.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1215 | Chr.14:23134948 - 23167798 on Build GRCh38 | Insertion | SLC7A8 |
nsv564040 | Chr.14:23126512 - 23163297 on Build GRCh38 | Loss | SLC7A8 |
More Information
Additional Information:
For this assay, SNP(s) [rs74667093] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |