Assay Details
Target Gene Details
Entrez Gene ID: | 51527 |
Gene Name: | GSK3B interacting protein |
Gene Aliases: |
C14orf129, HSPC210 |
Location: |
Chr.14:96363452-96387290 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GSKIP | NM_001271904.1 | NP_001258833.1 | ||
NM_001271905.1 | NP_001258834.1 | |||
NM_001271906.1 | NP_001258835.1 | |||
NM_016472.4 | NP_057556.2 | |||
XM_017021365.1 | XP_016876854.1 | |||
AF151044.1 | AAF36130.1 | |||
AK000796.1 | BAA91380.1 | |||
AK094654.1 | ||||
AK314423.1 | ||||
DA488110.1 | ||||
HY019704.1 | ||||
HY051680.1 | ||||
HY108584.1 |